FitMe™ Lab Jackets
Catalog # 76582-128
Supplier: Dukal
Specifications
- Clothing Size:XXL
- Closure type:Press studs / snaps
- Color:Lavender
- Fabric:Premium SMS
- Sleeve or leg length:Long
- Cat. no.:76582-128
Specifications
About this item
FitMe™ lab jackets feature a unique adjustable waistband design that provides a true custom fit.
- Knit cuffs and collar
- Snap front closure
- Premium SMS material
- 2 hip pockets, 1 chest pocket
- Washable and autoclavable up to 5 times
Available in a wide variety of colors, FitMe™ lab jackets are manufactured from premium SMS material, and are supplied non sterile.
Packaging: Lab jackets are packaged 10 per bag, 5 bags per case.