Eisco® Ring and Strings for Force Table
Catalog # 470346-968
Supplier: Wards
Specifications
- Description:Replacement ring and strings for 40 cm force tables
- Cat. no.:470346-968
Specifications
About this item
Replacement ring and strings for force table.
- Consists of one top ring and four 12" strings with terminal rings for attaching masses
Place center ring over central hub of table and drape radial strings over four pulleys attached 90 degrees from each other. Replacement part for Force Table part number 470014-356.