Order Entry
Canada
ContactUsLinkComponent
 
General Purpose Unstirred Digital Water Bath Range, 230 V, Grant Instruments
 
undefined
General Purpose Unstirred Digital Water Bath Range, 230 V, Grant Instruments

 

  • Catalog No:
  • 89500-388
  • 89500-386
  • 89500-392
  • 89500-390
  • Temperature range:
  • ambient +5 to 95°C
  • ambient +5 to 95°C
  • ambient +5 to 95°C
  • ambient +5 to 95°C
  • Temperature resolution:
  • 0.5 °C (display and setting)
  • 0.5 °C (display and setting)
  • 0.5 °C (display and setting)
  • 0.5 °C (display and setting)
  • Temperature stability:
  • 0.5 °C (DIN 12876) @ 70°C
  • 0.5 °C (DIN 12876) @ 70°C
  • 0.5 °C (DIN 12876) @ 70°C
  • 0.5 °C (DIN 12876) @ 70°C

 

Frequently Bought Together