ContactUsLinkComponent
Staticide® 7255 Polyester Swab with ESD Handle
Staticide® 7255 Polyester Swab with ESD Handle
Catalog # 77200-560
Supplier:  ACL Staticide
undefined
Staticide® 7255 Polyester Swab with ESD Handle
Catalog # 77200-560
Supplier:  ACL Staticide
Supplier Number:  7255
Specifications
  • Length:
    2.75"
  • Material:
    Polyester knit
  • Environmentally Preferable:
  • Cat. no.:
    77200-560
  • Tip Style:
    Round
Specifications
About this item
Robust and reliable knit swab for cleanroom applications that require static control.
  • Ultra-low particle; lint-free
  • Free of silicon, amides, and DOP contamination
  • Blue polypropylene handles have a resistance of 10⁸ to 10¹° ohms
  • Class 10 laundered and packaged
  • ISO Class 5 suitable
The 7255 polyester swab features a non-abrasive, double-layer polyester knit tip. The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.