Staticide® Polyurethane Foam Swab with ESD Handle
Catalog # 77200-556
Supplier: ACL Staticide
Specifications
- Length:2.5"
- Material:Medical-grade foam
- Environmentally Preferable:
- Cat. no.:77200-556
- Tip Style:Cone
Specifications
About this item
Medical-grade foam tipped swab with superior solvent capacity.
- High solvent capacity
- Non-abrasive medical grade foam
- Blue polypropylene handle has resistance of 10⁸ to 10¹° ohms
- Class 10 laundered and packaged
- ISO Class 4 - 7 compatible
The swab construction features ultra sonic welding, with no adhesives or binders utilized.
Packaging: Swabs are packaged 500 per bag, 2 bags per case.