Order Entry
Canada
ContactUsLinkComponent
Staticide® Low-Lint Wipes, Blue
Staticide® Low-Lint Wipes, Blue
Catalog # 77200-766
Supplier:  ACL Staticide
undefined
Staticide® Low-Lint Wipes, Blue
Catalog # 77200-766
Supplier:  ACL Staticide
Supplier Number:  LF99B

Specifications

  • Color:
    Blue
  • Material:
    Hydroentangled 45% polyester/55% cellulose
  • Cat. no.:
    77200-766
  • Packaging:
    300 wipes/bag, 6 bags/case
  • W×L:
    22.9×22.9 cm (9×9")

Specifications

About this item

Exceptional all-purpose performance that is suitable for tough jobs in any workshop or manufacturing facility.

  • 55% cellulose/45% polyester hydroentangled non-woven wipe
  • Maintains strength in wet applications
  • Compatible with ISO Class 6+ (Class 1000) cleanrooms
  • Excellent choice for general industrial manufacturing use
  • 1/4 fold packaging

Blue color shows liquids easier than white wipes to assure spills are lifted.