ContactUsLinkComponent
Staticide® 8120 Economy Heel Grounder
Staticide® 8120 Economy Heel Grounder
Catalog # 77200-598
Supplier:  ACL Staticide
undefined
Staticide® 8120 Economy Heel Grounder
Catalog # 77200-598
Supplier:  ACL Staticide
Supplier Number:  8120
Specifications
  • Color:
    Black
  • Description:
    Economy heel grounder
  • Cat. no.:
    77200-598
Specifications
About this item
The economy heel grounder is a cost-effective solution offering form-fitting comfort.
  • Two-layer heel rubber
  • Fits a variety of footwear
  • 1-meg ohm resistor
  • Black conductive exterior: 10⁴ ohms
This economy heel grounder features a conductive ground ribbon measuring 14.5×0.05", and can be secured with an adjustable hook and loop fabric with a D-ring tie-down. It is specifically designed for use by grounding employees on the move in electronic manufacturing areas.