ContactUsLinkComponent
Staticide® Common Point Ground Cord for ESD Mats, 10 mm Stud with Dual Port
Staticide® Common Point Ground Cord for ESD Mats, 10 mm Stud with Dual Port
  77200-570
undefined
Staticide® Common Point Ground Cord for ESD Mats, 10 mm Stud with Dual Port
  77200-570
 :  8091
  • Description:
    Common point ground cord for ESD mats
  • Cat. no.:
    77200-570
The 8091 common point mat ground cord snaps onto a groundable point on an ESD-safe mat.
  • 10' coated high-strength tensile wire
  • 1-meg ohm resistor
  • 10mm male stud connection to mats
  • Ring-style termination for connecting to ground point
  • Equipped with two banana jack plug-ins
This ground point features two plug-in banana jacks for wrist straps while connecting mats to a ground source.