ContactUsLinkComponent
Staticide® 8102 Economy Wriststrap Cord, 10 mm to Banana
Staticide® 8102 Economy Wriststrap Cord, 10 mm to Banana
  77200-576
undefined
Staticide® 8102 Economy Wriststrap Cord, 10 mm to Banana
  77200-576
 :  8100
  • Color:
    Dark blue
  • Description:
    Economy wriststrap cord, 10 mm to banana
  • Environmentally Preferable:
  • Cat. no.:
    77200-576
Economical grounding cord with 1-meg ohm resistor.
  • 6 foot cord with flexible strain relief assembly to prevent breakage
  • Recommended for use with 10 mm wriststraps
  • 10 mm socket-to-banana
  • Includes alligator clip
  • Anti-static nylon
The 8102 Economy wriststrap cord is equipeed with a 10 mm female socket. Fits to single-wires wrist straps with 10 mm male stud.
 : 6 cords per pack.