Staticide® ESD Traction Floor Mats
Supplier: ACL Staticide
New Product
Certificates
About this item
ACL Staticide conductive nitrile rubber ESD traction floor mats are ideal for environments that need slip-resistance and reliable static control.
- Resistance measures 10³ to 10⁵ ohms
- Single-layer 0.125" (38.1 mm) thickness
- Made in USA
ESD traction floor mats provide a high traction, slip-resistant surface for a variety of working environments.
Customers Who Bought This Also Bought