Orders LinkContactUsLinkComponent
Staticide® ESD Knit Gloves
Supplier:   ACL Staticide
Staticide® ESD Knit Gloves
Supplier:  ACL Staticide
Certificates
Staticide® ESD Knit Gloves
5 Options Available Below
Product Details & Documents
These ESD Knit gloves are ideal for inspection and assembly.
  • Seamless knit construction
  • Nylon conductive carbon fiber blend yarn
  • Low moisture absorption
  • GLK CAFE RGP: 10⁷ ohms
These micro-knit static control gloves offer superb comfort and durability. Ideal for inspection and assembly work. Ideal for inspection and assembly work when static control is required.
 : Ordered by case, six packs (36 pairs of gloves) per case.
 : Each pack contains six pairs.
Specifications for Product Family
Specifications
  • Catalog No:
  • 77200-752
  • 77200-750
  • 77200-748
  • 77200-764
  • 77200-746
  • Ambidextrous/Hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Nylon
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
Specifications
Product Family Options
Product Information
  • Glove size
    Glove color
    Length
    Thickness
    Availability
    Price
  • 77200-752
    Staticide ESD knit gloves with white rib
    Staticide ESD knit gloves with white rib
    Small
    White
    25.4 cm (10")
    13 gauge
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-S
    Specifications:
    More
    Specifications
    Cat. No.
    77200-752
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • 77200-750
    Staticide ESD knit gloves with green rib
    Staticide ESD knit gloves with green rib
    Medium
    White
    25.4 cm (10")
    13 gauge
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-M
    Specifications:
    More
    Specifications
    Cat. No.
    77200-750
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • 77200-748
    Staticide ESD knit gloves with brown rib
    Staticide ESD knit gloves with brown rib
    Large
    White
    25.4 cm (10")
    13 gauge
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-L
    Specifications:
    More
    Specifications
    Cat. No.
    77200-748
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • 77200-764
    Staticide ESD knit gloves with yellow rib
    Staticide ESD knit gloves with yellow rib
    X-Large
    White
    25.4 cm (10")
    13 gauge
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $214.49
    $214.49
    Case of 500
       $214.49
    Supplier Number: GLK-XL
    Specifications:
    More
    Specifications
    Cat. No.
    77200-764
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile
  • 77200-746
    Staticide ESD knit gloves with gray rib
    Staticide ESD knit gloves with gray rib
    2X-Large
    White
    25.4 cm (10")
    13 gauge
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $216.75
    $216.75
    Case of 500
       $216.75
    Supplier Number: GLK-2XL
    Specifications:
    More
    Specifications
    Cat. No.
    77200-746
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
    Powdered/Powder-free
    Powder-free
    Material
    Nylon
    Sterility
    Non sterile