Code Corporation Code Reader™ 5200 Barcode Scanner
Seamlessly connect your operations with the CR5200. It reads barcodes in any orientation, plus offers hands-free operation to expedite workflows.
- Rotational tolerance ±180°
- Field of view 51° horizontal by 39.4° vertical
- CMOS 1.2 megapixel sensor
- Dark gray color
- Cabled scanner connectivity (6' cable length)
- No battery
Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies. Quickly read barcodes on mobile device screens. Compact and adjustable for any reading direction. Provides user feedback with LED and audible tone. Features extended read ranges.
Model 176696 features age verification when scanning ID cards.
: Includes (1) CR5200; (1) USB cable.
Seamlessly connect your operations with the CR5200. It reads barcodes in any orientation, plus offers hands-free operation to expedite workflows.
- Rotational tolerance ±180°
- Field of view 51° horizontal by 39.4° vertical
- CMOS 1.2 megapixel sensor
- Dark gray color
- Cabled scanner connectivity (6' cable length)
- No battery
Get high-speed, omnidirectional reading of 1D and 2D barcode symbologies. Quickly read barcodes on mobile device screens. Compact and adjustable for any reading direction. Provides user feedback with LED and audible tone. Features extended read ranges.
Model 176696 features age verification when scanning ID cards.
: Includes (1) CR5200; (1) USB cable.
- Catalog No:
- 77282-158
- 77282-160
- Interfaces:
- RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
- RS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]
- Sensor type:
- CMOS 1.2 MP
- CMOS 1.2 MP
Product Family Options
- DescriptionDimensions
- : 176695
Specifications
Cat. No.77282-158InterfacesRS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]Sensor typeCMOS 1.2 MP - CR5200 Fixed barcode scanner with age verification3W×3.5D×5.5"HEach$1,146.88: 176696
Specifications
Cat. No.77282-160InterfacesRS232; USB 2.0 (generic HID, HID keyboard, virtual COM port) [reader/scanner unit]Sensor typeCMOS 1.2 MP