About this item
Powerful benchtop centrifugation for clinical use in stat laboratories, medical clinics and offices, veterinarian offices, hospitals and education.
- Programmable with memory for up to 8 programs
- Adjustable time (30 seconds to 60 minutes or continuous)
- Rapid acceleration
- Gentle electronic braking (deceleration time of <60 seconds)
- Digitally controlled brushless motor means accurate speed and no motor maintenance
This clinical centrifuge has an easy to read digital back-lit display, it features tool-less rotor removal for convenient loading and cleaning. The clear lid with viewing porthole allows non-contact speed verification (with non-contact tachometer). Unit has a compact footprint for benchtop use. Accomodates 3 to 10 ml blood collection tubes, 5 ml microcentrifuge tubes, and 15 ml conical centrifuge tubes.
Built-in safety features include automatic shutoff if lid is open, if unit is jammed or out of balance. A manual lid unlock allows users to open lid in case of power failure and prevents loss of samples.
Maximum relative humidity: Non-condensing, 80% for temperatures up to 31 °C decreasing linearly to 50% R.H at 40 °C.
Specifications
- Catalog No:
- 470353-782
- 470353-784
- Power:
- Approximately 93 W
- Approximately 93 W