MaxiChem® Cut™ Nitrile Blend Coated Gloves with TriTech™ Liner and Non-Slip Grip on Palm and Fingers
Supplier: PIP
New Product
Certificates
About this item
MaxiChem® Cut™ cut-resistant nitrile chemical gloves.
- Chemical resistance delivered through ATG® LiquiTech® technology platform
- Triple-layered TriTech™ technology for added protection, performance, and comfort
- Tailored cuff keeps fingers at the tip of the glove to maximize dexterity
- Micro-cup non-slip grip finish allows for a controlled and superior grip in oily and wet applications
- Non-slip grip is only applied where needed (palm area) to ensure flexibility isn't compromised
Chemical cut gloves feature innovative fibers and base ingredients developed into unique, high-performance yarns and fibers that offer cut protection while maintaining high levels of comfort. An ultra-lightweight synthetic coating, combined with an industry leading super-light seamless knit liner, provides excellent comfort, sensitivity, fit, and dexterity.
Certifications: EN 374 results: JKLMNO. ANSI Cut Level: A2. EN 388 2016 results: 4321BX.
Specifications
- Catalog No:
- 77320-762
- 77320-760
- 77320-764
- 77320-768
- 77320-766
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile