Magnetic Wands
470353-638
- Description:Magnetic wands
- Cat. no.:470353-638
Colorful wands to explore magnetism.
- Six wands in assorted colors
- Designed for simple magnetic activities
- Magnet encased in plastic
This easy-to-hold plastic wand contains a ceramic magnet. Use for various activities to learn what items are attracted to magnets. Each wand measures 8" long by 1" wide. Sold as a set of six magnets in assorted colors.