Anti-MAP2 Mouse Monoclonal Antibody [clone: 4H5]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (4H5) antibody to MAP2 for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse and Bovine.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:10000, ICC/IF: 1:1000, IHC: 1:100
Type: Primary
Antigen: MAP2
Clonality: Monoclonal
Clone: 4H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse;Bovine
Specifications
- Catalog No:
- 77079-414
- 77073-944
- Antigen symbol:
- MAP2
- MAP2
- Antigen name:
- DKFZp686I2148
- DKFZp686I2148
- Antigen synonyms:
- MAP2A|Microtubule-associated protein 2|MAP dendrite specific|MAP2C|MAP2|MAP-2|MAP 2|MAP2B|Microtubule associated protein 2
- MAP2A|Microtubule-associated protein 2|MAP dendrite specific|MAP2C|MAP2|MAP-2|MAP 2|MAP2B|Microtubule associated protein 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4H5
- 4H5
- Reactivity:
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P11137
- UniprotID# P11137
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Purified full-length bovine brain MAP2
- Purified full-length bovine brain MAP2
- Form:
- Liquid
- Liquid
- Molecular weight:
- ~280 kDa (by SDS-PAGE)
- ~280 kDa (by SDS-PAGE)
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC
Specifications
Frequently Bought Together