Anti-UROD Goat Polyclonal Antibody
Catalog # 76953-714
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DCUP_HUMAN
- Antigen symbol:UROD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:URO-D|URO D|Uroporphyrinogen decarboxylase|UPD|UROD|PCT|Uroporphyrinogen III decarboxylase
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:KARECVGKTVTLQGN
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human UROD (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:76953-714
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to UROD for ELISA, WB and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: UROD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human