Anti-Proteasome 20S beta 7 Goat Polyclonal Antibody
76958-590
- Antibody type:Primary
- Antigen name:Macropain chain Z
- Antigen symbol:Proteasome 20S beta 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Proteasome beta 7 subunit|Multicatalytic endopeptidase complex chain Z|Proteasome subunit beta 7|Proteasome subunit alpha|Proteasome subunit beta type-7|Proteasome subunit Z|Proteasome (prosome macropain) subunit beta type 7|Proteasome 20S beta 7|Proteasome catalytic subunit 2
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-RNAVLEADFAKR
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human PSMB7 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:76958-590
- Packaging:Plastic vial
Goat polyclonal antibody to Proteasome 20S beta 7 for ELISA and WB with samples derived from Human and Mouse.
- Validated Applications: ELISA, W
Type: Primary
Antigen: Proteasome 20S beta 7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse