Anti-Aconitase 2 Goat Polyclonal Antibody (Biotin)
Catalog # 77080-888
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:ACO 2
- Antigen symbol:Aconitase 2
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:ACO2|Aconitate hydratase|Aconitase 2 mitochondrial|Aconitase2|Aconitate hydratase mitochondrial|ACONM|Aconitase|Aconitase 2|ACON_HUMAN
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CQDTYQHPPKDSSGQH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human ACO2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77080-888
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to ACO2 (Biotin) for ELISA and WB with samples derived from Human, Mouse, Rat and Porcine.
- Validated Applications: ELISA, W
Type: Primary
Antigen: Aconitase 2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat;Porcine