Anti-ACAT1 Goat Polyclonal Antibody
Catalog # 77066-858
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:ACAT
- Antigen symbol:ACAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Acetoacetyl CoA thiolase|acetyl-coa acetyltransferase precursor, mitochondrial|Acetoacetyl-CoA thiolase|Acetyl-CoA acetyltransferase|Acetyl Coenzyme A acetyltransferase 1|Acetyl CoA acetyltransferase, mitochondrial|ACAT 1|acetoacetyl Coenzyme A thiolase|ACAT1
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KRVDFSKVPKLKT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human ACAT1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77066-858
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to ACAT1 for ELISA, WB and IF with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, WB, I
Type: Primary
Antigen: ACAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat