Anti-IRE1 Goat Polyclonal Antibody
Catalog # 77073-860
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Endoplasmic reticulum (ER) to nucleus signalling 1
- Antigen symbol:IRE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:ERN1_HUMAN|Endoribonuclease|Endoplasmic reticulum-to-nucleus signaling 1|hIRE1p|ERN 1|Endoplasmic reticulum to nucleus signaling 1|hIRE 1p|ER to nucleus signaling 1|Ern1
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-PNAHGKIKAMISD
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human ERN1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77073-860
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to IRE1 for ELISA, WB and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, WB, Flow Cytometr
Type: Primary
Antigen: IRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human