Anti-TRPV5 Goat Polyclonal Antibody
Catalog # 77074-144
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Calcium transport protein 2
- Antigen symbol:TRPV5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:Epithelial calcium channel 1|ECAC 1|ECaC|Calcium transporter 2|CAT 2|Osm 9 like TRP channel 3|ECaC1|CAT2|Osm-9-like TRP channel 3
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-SHRGWEILRQNT
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human TRPV5 (internal region)
- Tested applications:ICC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77074-144
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to TRPV5 for ELISA, WB, ICC and IF with samples derived from Human.
- Validated Applications: ELISA, WB, ICC, I
Type: Primary
Antigen: TRPV5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human