Anti-RNF213 Goat Polyclonal Antibody
Catalog # 77076-696
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:ALK lymphoma oligomerization partner on chromosome 17
- Antigen symbol:RNF213
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen synonyms:ALO17_HUMAN|KIAA1618|E3 ubiquitin-protein ligase RNF213|C17orf27|Chromosome 17 open reading frame 27|MGC46622|MGC9929|KIAA1554|FLJ13051
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KTAAVLKWNRE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human RNF213 (C terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77076-696
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to RNF213 for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IH
Type: Primary
Antigen: RNF213
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human