Anti-ZNF217 Goat Polyclonal Antibody
77066-294
- Antibody type:Primary
- Antigen name:ZABC 1
- Antigen symbol:ZNF217
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen synonyms:ZN217_HUMAN|ZABC1|ZN 217|ZNF217|Zinc finger protein 217|ZN217|ZNF 217
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KNAADDSADKVNKN
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human ZNF217 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77066-294
- Packaging:Plastic vial
Goat polyclonal antibody to ZNF217 for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IH
Type: Primary
Antigen: ZNF217
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human