Anti-14-3-3 sigma Goat Polyclonal Antibody
Catalog # 77077-766
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:14 3 3 protein sigma
- Antigen symbol:14-3-3 sigma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:MGC143283|Mkrn3|1433S_HUMAN|14-3-3 sigma|Er|HME 1|Epithelial cell marker protein 1|14-3-3 protein sigma|HME1
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-DLHTLSEDSYKDST
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human SFN (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77077-766
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to 14-3-3 sigma for ELISA, WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: 14-3-3 sigma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat