Anti-FEM1A Goat Polyclonal Antibody
77067-990
- Antibody type:Primary
- Antigen name:AW611390
- Antigen symbol:FEM1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:FEM1a-A|Fem-1 homolog a (C. elegans)|DKFZp762M136|Fem1aa|FEM1-alpha-A|Fem1 homolog a|FEM1-alpha|EPRAP|FEM1A
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-GFIPEDLEAFIELH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human FEM1A (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77067-990
- Packaging:Plastic vial
Goat polyclonal antibody to FEM1A for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, W
Type: Primary
Antigen: FEM1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human