Anti-DKK4 Goat Polyclonal Antibody
Catalog # 77062-664
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Dickkopf 4
- Antigen symbol:DKK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:DKK 4|Dickkopf homolog 4|Dickkopf related protein 4|Dkk-4|Dickkopf gene 4|Dickkopf homolog 4 (Xenopus laevis)|Dickkopf-related protein 4 short form|Dickkopf-4|Dickkopf4
- Storage buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:RQHARLRVCQKIEKL
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human DKK4 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77062-664
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to DKK4 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, W
Type: Primary
Antigen: DKK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human