Anti-Uridine Phosphorylase 1 Rabbit Monoclonal Antibody [clone: ARC2531]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2531] antibody to Uridine Phosphorylase 1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Uridine Phosphorylase 1
Clonality: Monoclonal
Clone: ARC2531
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77223-556
- Antibody type:
- Primary
Specifications
Related Information
Frequently Bought Together