Anti-B3GNT1 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to B3GNT1 for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: B3GNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77222-210
- Antigen symbol:
- B3GNT1
- Antigen name:
- 3-galactosyltransferase 7
- Antigen synonyms:
- B3GN2_HUMAN|b3Gal-T7|3-N-acetylglucosaminyltransferase 2|B3GNT1|3-GalTase 7|3-N-acetylglucosaminyltransferase 1|3-Gn-T1|B3GNT|3-Gn-T2
- Antibody type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9NY97
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 29-240 of human B3GNT2 (NP_006568.2).
- Sequence:
- KSSSQEKNGKGEVIIPKEKFWKISTPPEAYWNREQEKLNRQYNPILSMLTNQTGEAGRLSNISHLNYCEPDLRVTSVVTGFNNLPDRFKDFLLYLRCRNYSLLIDQPDKCAKKPFLLLAIKSLTPHFARRQAIRESWGQESNAGNQTVVRVFLLGQTPPEDNHPDLSDMLKFESEKHQDILMWNYRDTFFNLSLKEVLFLRWVSTSCPDTEF
- Form:
- Liquid
- Molecular weight:
- 46 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together