Anti-Serine/threonine-protein kinase 4/MST-1 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Serine/threonine-protein kinase 4/MST-1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:100, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Serine / threonine-protein kinase 4 / MST-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77222-162
- Antibody type:
- Primary
Specifications
Related Information
Frequently Bought Together