Anti-CDK1 (phospho Tyr15) Rabbit Monoclonal Antibody [clone: ARC55443]
Rabbit monoclonal [ARC55443] antibody to CDK1 (phospho Tyr15) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: CDK1
Clonality: Monoclonal
Clone: ARC55443
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77220-866
- Antibody type:
- Primary
Frequently Bought Together