Anti-PHOX2B Rabbit Monoclonal Antibody [clone: ARC1420]
Rabbit monoclonal [ARC1420] antibody to PHOX2B for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: PHOX2B
Clonality: Monoclonal
Clone: ARC1420
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77220-338
- Antigen symbol:
- PHOX2B
- Antigen name:
- NBLST2
- Antigen synonyms:
- PHOX 2B|Neuroblastoma paired type homeobox protein|PHOX 2B homeodomain protein|Paired-like homeobox 2B|Paired mesoderm homeobox protein 2B|Paired mesoderm homeobox 2b|Paired like homeobox 2b|NBPhox|Neuroblastoma Phox
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1420
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q99453
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 215-314 of human PHOX2B (Q99453).
- Sequence:
- GGGPSPAGAPGAAGPGGPGGEPGKGGAAAAAAAAAAAAAAAAAAAAGGLAAAGGPGQGWAPGPGPITSIPDSLGGPFASVLSSLQRPNGAKAALVKSSMF
- Form:
- Liquid
- Molecular weight:
- 35 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Frequently Bought Together