Anti-PELP1 Rabbit Monoclonal Antibody [clone: ARC1387]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1387] antibody to PELP1 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: PELP1
Clonality: Monoclonal
Clone: ARC1387
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77219-394
- Antigen symbol:
- PELP1
- Antigen name:
- glutamic acid- and leucine-rich protein 1
- Antigen synonyms:
- HMX3|PELP1|PELP1 proline glutamic acid leucine rich protein 1|proline and glutamic acid rich nuclear protein|MNAR|Modulator of nongenomic activity of estrogen receptor|PELP1_HUMAN|Modulator of non-genomic activity of estrogen receptor|P160
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1387
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q8IZL8
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 1000-1100 of human PELP1 (Q8IZL8).
- Sequence:
- EPEPGLLLEVEEPGTEEERGADTAPTLAPEALPSQGEVEREGESPAAGPPPQELVEEEPSAPPTLLEEETEDGSDKVQPPPETPAEEEMETETEAEALQEK
- Form:
- Liquid
- Molecular weight:
- 160 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together