Anti-PI 3 Kinase p55 gamma Rabbit Monoclonal Antibody [clone: ARC0320]
Rabbit monoclonal [ARC0320] antibody to PI 3 Kinase p55 gamma for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: PI 3 Kinase p55 gamma
Clonality: Monoclonal
Clone: ARC0320
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77218-650
- Antigen symbol:
- PI 3 Kinase p55 gamma
- Antigen name:
- DKFZp686P05226
- Antigen synonyms:
- Phosphatidylinositol 3 kinase regulatory subunit polypeptide 3|p55PIK|Phosphatidylinositol 3 kinase regulatory subunit gamma|FLJ41892|p55|P55G_HUMAN|OTTHUMP00000009783|OTTHUMP00000009786|p55 gamma
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0320
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q92569
- Isotype:
- IgG
- Western blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 362-461 of human PIK3R3/p55PIK (Q92569).
- Sequence:
- DINRVQAEDLLYGKPDGAFLIRESSKKGCYACSVVADGEVKHCVIYSTARGYGFAEPYNLYSSLKELVLHYQQTSLVQHNDSLNVRLAYPVHAQMPSLCR
- Form:
- Liquid
- Molecular weight:
- 52 kDa
- Purification:
- Affinity purification
- Storage buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
Frequently Bought Together