Anti-AHA1 Mouse Monoclonal Antibody [clone: 4H9-D7]
Catalog # 77172-574
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Activator of 90 kDa heat shock protein ATPase homolog 1
- Antigen symbol:AHA1
- Clonality:Monoclonal
- Clone:4H9-D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O95433
- Antigen synonyms:HSPC322|AHA1|C14orf3|Activator of heat shock 90kDa protein ATPase homolog 1|p38|AHSA 1|AHSA1_HUMAN|Ahsa1|AHA 1
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.2, with 50% Glycerol and 0.09% Sodium Azide
- Molecular weight:~38 kDa (can run up to 45 kDa on SDS-PAGE)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human Aha1, his tagged.
- Tested applications:ICC/IF
- Purification:Protein G purification
- Cat. no.:77172-574
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [4H9-D7] antibody to AHA1 for WB, ICC/IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ICC/IF, ELISA
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100
Type: Primary
Antigen: AHA1
Clonality: Monoclonal
Clone: 4H9-D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse