Anti-Methylglyoxal Mouse Monoclonal Antibody [clone: 9E7]
Catalog # 77172-218
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:2 oxo propanal
- Antigen symbol:Methylglyoxal
- Clonality:Monoclonal
- Clone:9E7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Species independent
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen synonyms:pyruvaldehyde|MG|Methylglyoxal
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH)
- Tested applications:ICC/IF, IHC, FACS
- Purification:Protein G purification
- Cat. no.:77172-218
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [9E7] antibody to Methylglyoxal for WB, ICC/IF, IHC, FACS, Flow Cytometry and ELISA with samples derived from Species Independent.
- Validated Applications: WB, ICC/IF, IHC, FACS, Flow Cytometry, ELISA
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:50, FACS: 1:50, FCM: 1:50, ELISA: 1:1,000, IHC: 1:50
Type: Primary
Antigen: Methylglyoxal
Clonality: Monoclonal
Clone: 9E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent