Anti-Horse IgG H&L Rabbit Polyclonal Antibody (Biotin)
Catalog # 76835-166
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Secondary
- Antigen symbol:Horse IgG H&L
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Horse
- Western blot:Yes
- Size:1 mg
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in Phosphate Buffered Saline with 0.2% BSA and 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Store at 4 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. no.:76835-166
- Packaging:Plastic vial
Specifications
About this item
Rabbit anti-horse IgG H&L (Biotin) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Horse.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:1,000 to 1:30,000; ELISA (capture antibody): 1:100 to 1:500, ICC: 1:200 to 1:2,000, IHC: 1:200 to 1:2,000, WB: 1:1,000 to 1:30,000
Type: Secondary
Antigen: Horse IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Horse