Anti-STAT3 Rabbit Recombinant Antibody [clone: RM465]
Catalog # 76834-362
Supplier: Antibodies.com
Specifications
- Antibody type:Primary
- Antigen name:1110034C02Rik
- Antigen symbol:STAT3
- Clonality:Recombinant
- Clone:RM465
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:UniprotID# P40763
- Antigen synonyms:AW109958|Acute-phase response factor|ADMIO|Acute Phase Response Factor|HIES|FLJ20882|APRF|MGC16063|DNA binding protein APRF
- Storage buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the N terminus of human STAT3.
- Tested applications:IHC
- Purification:Protein A chromatography
- Cat. no.:76834-362
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [RM465] antibody to STAT3 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
- Recommended Dilutions: IHC: 1:50 to 1:200, WB: 1:1,000 to 1:2,000
Type: Primary
Antigen: STAT3
Clonality: Recombinant
Clone: RM465
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human