Anti-Phosphoglucomutase 5 Rabbit Recombinant Antibody [clone: PGM5/7005R]
Catalog # 76956-598
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Phosphoglucomutase 5
- Antigen symbol:Phosphoglucomutase 5
- Clonality:Recombinant
- Clone:PGM5/7005R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Rabbit,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15124
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:56 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeletal fraction of the human myometrium tissue extract.
- Purification:Protein A/G chromatography
- Cat. no.:76956-598
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [PGM5/7005R] antibody to Phosphoglucomutase 5 for ELISA, IF and WB with samples derived from Human, Bovine, Mouse, Rat and Rabbit.
- Validated Applications: ELISA, IF, WB
- Recommended Dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: Phosphoglucomutase 5
Clonality: Recombinant
Clone: PGM5/7005R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG2a kappa
Reactivity: Human;Bovine;Mouse;Rat;Rabbit