Anti-PRMT7 Mouse Monoclonal Antibody [clone: PCRP-PRMT7-1A7]
Catalog # 76966-086
Supplier: Antibodies.com
Specifications
- Antibody type:Primary
- Antigen name:ANM7_HUMAN
- Antigen symbol:PRMT7
- Clonality:Monoclonal
- Clone:PCRP-PRMT7-1A7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NVM4
- Antigen synonyms:KIAA1933|Protein arginine N-methyltransferase 7|Myelin basic protein arginine N methyltransferase|Histone-arginine N-methyltransferase PRMT7|OTTHUMP00000174863|FLJ10640|[Myelin basic protein]-arginine N-methyltransferase PRMT7|Protein arginine methyltransferase 7
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:78.46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PRMT7 protein.
- Purification:Protein A/G chromatography
- Cat. no.:76966-086
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-PRMT7-1A7] antibody to PRMT7 for IP, WB, IF and Flow Cytometry with samples derived from Human.
- Validated Applications: IP, WB, IF, Flow Cytometry
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: PRMT7
Clonality: Monoclonal
Clone: PCRP-PRMT7-1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human