Anti-PBX1 Mouse Monoclonal Antibody [clone: PCRP-PBX1-3C8]
76964-590
- Antibody type:Primary
- Antigen name:DKFZp686B09108
- Antigen symbol:PBX1
- Clonality:Monoclonal
- Clone:PCRP-PBX1-3C8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P40424
- Antigen synonyms:MGC126627|Homeobox protein PRL|Homeobox protein PBX1|Homeobox protein PBX 1|PBX1_HUMAN|Homeo box protein PBX1|Pre B cell leukemia transcription factor 1|Pre B cell leukemia homeobox 1|Homeo box protein PRL
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:86.1 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PBX1 protein.
- Purification:Protein A/G chromatography
- Cat. no.:76964-590
- Packaging:Plastic vial
Mouse monoclonal [PCRP-PBX1-3C8] antibody to PBX1 for ELISA, IP, Flow Cytometry and IF with samples derived from Human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: PBX1
Clonality: Monoclonal
Clone: PCRP-PBX1-3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human