Anti-TFIIB Mouse Monoclonal Antibody [clone: PCRP-GTF2B-1D1]
76949-376
- Antibody type:Primary
- Antigen name:General transcription factor IIB
- Antigen symbol:TFIIB
- Clonality:Monoclonal
- Clone:PCRP-GTF2B-1D1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q00403
- Antigen synonyms:Transcription initiation factor IIB|RNA polymerase II transcription factor IIB|TF IIB|gtf2b|S300-II|S300 II|TF2B_HUMAN|General transcription factor TFIIB
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:34.8 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TFIIB protein.
- Purification:Protein A/G chromatography
- Cat. no.:76949-376
- Packaging:Plastic vial
Mouse monoclonal [PCRP-GTF2B-1D1] antibody to TFIIB for ELISA, Flow Cytometry and IP with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry, IP
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IP: 1 to 2µg / 100 to 500µg proteins
Type: Primary
Antigen: TFIIB
Clonality: Monoclonal
Clone: PCRP-GTF2B-1D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human