Anti-Estrogen Receptor alpha Rabbit Recombinant Antibody [clone: ESR1/4039R]
Catalog # 76963-410
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686N23123
- Antigen symbol:Estrogen Receptor alpha
- Clonality:Recombinant
- Clone:ESR1/4039R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P03372
- Antigen synonyms:Estradiol receptor|ER|Era|ER alpha|ESRA|ER-alpha|ESR|ESR1|ESR1_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular weight:~67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the C terminus of human Estrogen Receptor alpha.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. no.:76963-410
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [ESR1/4039R] antibody to Estrogen Receptor alpha for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Estrogen Receptor alpha
Clonality: Recombinant
Clone: ESR1/4039R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human