Anti-BCL11A Mouse Monoclonal Antibody [clone: PCRP-BCL11A-1H3]
76950-996
- Antibody type:Primary
- Cat. no.:76950-996
Mouse monoclonal [PCRP-BCL11A-1H3] antibody to BCL11A for ELISA, IP, Flow Cytometry and IF with samples derived from Human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: BCL11A
Clonality: Monoclonal
Clone: PCRP-BCL11A-1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human