Anti-Haptoglobin Mouse Monoclonal Antibody [clone: HP/3835]
Catalog # 76951-466
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Cat. no.:76951-466
Specifications
About this item
Mouse monoclonal [HP/3835] antibody to Haptoglobin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Haptoglobin
Clonality: Monoclonal
Clone: HP/3835
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human