Anti-CD15 Mouse Monoclonal Antibody [clone: W6D3]
76957-090
- Antibody type:Primary
- Cat. no.:76957-090
Mouse monoclonal [W6D3] antibody to CD15 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: W6D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human