Anti-CD30-L Rabbit Recombinant Antibody [clone: DM201]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant rabbit monoclonal [DM201] antibody to CD30-L for ELISA and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: CD30-L
Clonality: Recombinant
Clone: DM201
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77186-670
- 77186-672
- Antigen symbol:
- CD30-L
- CD30-L
- Antigen name:
- CD153
- CD153
- Antigen synonyms:
- CD30 antigen ligand|CD30-L|CD30 L|CD30 ligand|MGC138144|CD30LG|CD30L|TNFL8_HUMAN|CD153 antigen
- CD30 antigen ligand|CD30-L|CD30 L|CD30 ligand|MGC138144|CD30LG|CD30L|TNFL8_HUMAN|CD153 antigen
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM201
- DM201
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P32971
- UniprotID# P32971
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together