Anti-NRG1 Rabbit Recombinant Antibody [clone: DM209]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant rabbit monoclonal [DM209] antibody to NRG1 for ELISA and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: NRG1
Clonality: Recombinant
Clone: DM209
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77186-638
- 77186-640
- Antigen symbol:
- NRG1
- NRG1
- Antigen name:
- Acetylcholine receptor-inducing activity
- Acetylcholine receptor-inducing activity
- Antigen synonyms:
- Heregulin|ARIA|Breast cancer cell differentiation factor p45|Acetylcholine receptor-inducing activity, CHICK, homolog of|Glial growth factor|heregulin, alpha (45kD, ERBB2 p185-activator)|GGF2|GGF|HGL
- Heregulin|ARIA|Breast cancer cell differentiation factor p45|Acetylcholine receptor-inducing activity, CHICK, homolog of|Glial growth factor|heregulin, alpha (45kD, ERBB2 p185-activator)|GGF2|GGF|HGL
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DM209
- DM209
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# Q02297
- UniprotID# Q02297
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
Specifications
Frequently Bought Together