Anti-CTLA4 Rabbit Recombinant Antibody [clone: DM51] (Biotin)
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Recombinant rabbit monoclonal [DM51] antibody to CTLA4 (Biotin) for ELISA and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: CTLA4
Clonality: Recombinant
Clone: DM51
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77182-288
- 77182-286
- Antigen symbol:
- CTLA4
- CTLA4
- Antigen name:
- ALPS5
- ALPS5
- Antigen synonyms:
- CD152 antigen|CD|CTLA 4|Celiac disease 3|CTLA-4|CD 152|CD152|CD152 isoform|CELIAC3
- CD152 antigen|CD|CTLA 4|Celiac disease 3|CTLA-4|CD 152|CD152|CD152 isoform|CELIAC3
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Biotin
- Biotin
- Clone:
- DM51
- DM51
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P16410
- UniprotID# P16410
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilized
- Lyophilized
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Lyophilized from sterile Phosphate Buffered Saline, pH 7.4. Normally 5 - 8% Trehalose is added as a protectant before lyophilization
- Storage temperature:
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
Specifications
Related Information
Frequently Bought Together