Anti-Periostin Mouse Monoclonal Antibody [clone: POSTN/3504]
Catalog # 77180-690
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody type:Primary
- Antigen name:Fasciclin-I like
- Antigen symbol:Periostin
- Clonality:Monoclonal
- Clone:POSTN/3504
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15063
- Antigen synonyms:Osteoblast specific factor|OSF2|OSF-2|Osteoblast specific factor 2|MGC119510|Osteoblast-specific factor 2|MGC119511|OSF 2|Osteoblast specific factor 2 (fasciclin I like)
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:84 kDa/74 kDa (Periostin) kDa/90 kDa (secreted glycoprotein)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 193-326) of human POSTN protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. no.:77180-690
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [POSTN/3504] antibody to Periostin for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: Periostin
Clonality: Monoclonal
Clone: POSTN/3504
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human